Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF500 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540435 Supplier Invitrogen™ Supplier No. PA540435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TGERPYKCLV CGKGFSDRSN FSTHQRVHTG EKPYPCPECG KRFSQSSSLV Sequence homology: Cow: 92%; Dog: 100%; Horse: 83%; Human: 100%; Mouse: 91%; Pig: 86%; Rabbit: 100%; Rat: 83%; Zebrafish: 82%.

ZNF500 is a new candidate transcription factor. It is derived from an annotated genomic sequence (NT_037887) using a gene prediction method.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF500
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF500
Gene Accession No. O60304
Gene Alias KIAA0557; LOC100069865; LOW QUALITY PROTEIN: zinc finger protein 500; zinc finger protein 500; Zinc finger protein with KRAB and SCAN domains 18; ZKSCAN18; ZNF500; ZSCAN50
Gene Symbols ZNF500
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF500.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26048
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.