Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF502 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF502 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF502 Polyclonal specifically detects ZNF502 in Human samples. It is validated for Western Blot.Specifications
ZNF502 | |
Polyclonal | |
Rabbit | |
NP_149987 | |
91392 | |
Synthetic peptide directed towards the N terminal of human ZNF502. Peptide sequence IDSSGIVVKRFQEDEYQDSTFEEKYACEGMKENSPREIAESCLFQEGGFG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
KOX17Zinc finger protein 191, lacks zinc finger domain, Retinoic acid suppression protein A, RSG-A, Zfp191, Zinc finger and SCAN domain-containing protein 3, zinc finger protein 24, zinc finger protein 24 (KOX 17), ZNF191, ZSCAN3Zinc finger protein KOX17 | |
ZNF502 | |
IgG | |
63 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title