Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF502 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556188 Supplier Invitrogen™ Supplier No. PA556188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FQEGGFGRIT FIHKEAPPEI ISQGYNFEKS LLLTSSLVTR LRVSTEESLH QWETSNIQTN DISDQSKCPT LCTQ Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 23%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF502
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF502
Gene Accession No. Q8TBZ5
Gene Alias zinc finger protein 502; ZNF502
Gene Symbols ZNF502
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF502. Recombinant protein control fragment (Product #RP-93641).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91392
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.