Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF503 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232132R
Description
ZNF503 Polyclonal antibody specifically detects ZNF503 in Human samples. It is validated for Western BlotSpecifications
ZNF503 | |
Polyclonal | |
Western Blot | |
KOX17Zinc finger protein 191, lacks zinc finger domain, Retinoic acid suppression protein A, RSG-A, Zfp191, Zinc finger and SCAN domain-containing protein 3, zinc finger protein 24, zinc finger protein 24 (KOX 17), ZNF191, ZSCAN3Zinc finger protein KOX17 | |
The immunogen this antibody was made to, maps to a region between residue 275 to 325 of human Zinc Finger Protein 503 using the numbering given in entry NP_116161.2 (GeneID 84858). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
84858 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction