Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF503 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555439 Supplier Invitrogen™ Supplier No. PA555439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPSSKLSSVA SNGGGAGGAG GGAAGDKDTK SGPLKLSDIG VEDKSSFKPY SKPGSDKKEP GGGGGGGGGG GGGGGGVSSE KSGFRVPSAT CQPFTPRTGS PSSSASACSP GGMLSSAGGA PEGKDDKKDT DVG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

The ZNF503 gene encodes zinc finger protein 503, a transcription factor involved in the regulation of developmental processes and cellular differentiation. ZNF503 contains zinc finger domains that enable it to bind DNA and modulate the transcription of specific target genes, influencing pathways critical for embryonic development, particularly in the neural and mesenchymal tissues. It plays a substantial role in the regulation of stem cell differentiation and has been implicated in various biological processes, including neuronal migration, immune response, and organogenesis. Alterations in ZNF503 expression have been linked to developmental anomalies and diseases such as cancers, where it affects tumor growth and progression by modulating gene networks involved in cell proliferation and differentiation. Research into ZNF503 focuses on understanding its complex regulatory functions and potential as a target for therapeutic interventions in developmental disorders and oncological applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF503
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF503
Gene Accession No. Q96F45
Gene Alias Nlz2; NOLZ1; NOLZ-1; Zinc finger protein 503; ZNF503
Gene Symbols ZNF503
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF503. Recombinant protein control fragment (Product #RP-102302).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.