Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF507 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178194H
Description
ZNF507 Polyclonal antibody specifically detects ZNF507 in Human samples. It is validated for Western BlotSpecifications
ZNF507 | |
Polyclonal | |
Western Blot | |
BMZF-1ZNF411, Bone marrow zinc finger 1, DNA-binding protein, FLJ90391, zinc finger protein 253, Zinc finger protein 411BMZF1 | |
The immunogen for this product maps to a region between residue 1 and 50 of human Zinc Finger Protein 507 using the numbering given in entry NP_055725.2 (GeneID 22847). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
22847 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction