Learn More
ZNF518A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303238AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May be involved in transcriptional regulation.Specifications
ZNF518A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6AHZ1 | |
ZNF518A | |
Between 1300 and 1350 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF518A | |
KIAA0335; ZNF518 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9849 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.