Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF521 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572983
Description
ZNF521 Polyclonal Antibody for ICC/IF, IHC (P)
The zinc finger protein 521 (ZNF521) is a transcription factor containing an N-terminal transcriptional repressor motif and 30 zinc finger domains. It plays a role in both erythroid cell and osteoblast differentiation during development, inhibiting the activities of early B-cell factor 1 (EBF1) in erythroid cells and Runx2 in osteoblast precursors. ZFP521 binds to both Runx2 and histone deacetylase 3 (HDAC3), promotes their association and antagonizes Runx2 transcriptional activity in a HDAC3-dependent manner, thereby regulating osteoblast differentiation, skeletal development, and bone homeostasis.
Specifications
| ZNF521 | |
| Polyclonal | |
| Unconjugated | |
| ZNF521 | |
| B930086A16Rik; early hematopoietic zinc finger protein; Ecotropic viral integration site 3 protein; EHZF; Evi3; LIP3; LYST-interacting protein 3; Zfp521; Zinc finger protein 521; Znf521 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 225207, 25925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 1.07 mg/mL | |
| PBS with 30% glycerol and 0.05% sodium azide | |
| Q6KAS7, Q96K83 | |
| Zfp521, ZNF521 | |
| A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 200-250). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction