Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF528 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541182 Supplier Invitrogen™ Supplier No. PA541182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LTSHQRIHTR ERPYGCSQCG KIFSQKSDLI RHRKTHTDEK PYKCNKCGTA Sequence homology: Cow: 79%; Dog: 79%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 77%; Rabbit: 83%; Rat: 79%.

ZNF528 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF528
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF528
Gene Accession No. Q3MIS6
Gene Alias KIAA1827; Zinc finger protein 528; ZNF528
Gene Symbols ZNF528
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF528.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84436
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.