Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF529 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF529 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF529 Polyclonal specifically detects ZNF529 in Human samples. It is validated for Western Blot.Specifications
ZNF529 | |
Polyclonal | |
Rabbit | |
NP_066002 | |
57711 | |
Synthetic peptide directed towards the N terminal of human ZNF529. Peptide sequence QMKIISENVPSYKTHESLTLPRRTHDSEKPYEYKEYEKVFSCDLEFDEYQ. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
KIAA1615, zinc finger protein 529 | |
ZNF529 | |
IgG | |
54 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title