Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF530 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120541 Supplier Invitrogen™ Supplier No. PA5120541
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-251MG, Mouse brain, Rat kidney. Immunogen sequence: MAAALRAPTQ QVFVAFEDVA IYFSQEEWEL LDEMQRLLYR DVMLENFAVM ASLGCWCGAV DEGTPSAESV SVEELSQGRT PKADTSTDKS HPCEICTPVL RDILQMIELH ASPCGQKLYL GGASRDFWMS SNLHQLQKLD NGEKLFKVDG DQASFMMNCR FHVSGKPFTF GEVGRDFSAT SGLLQHQVTP.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF530
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZNF530
Gene Accession No. Q6P9A1
Gene Alias KIAA1508; Zinc finger protein 530; ZNF530
Gene Symbols ZNF530
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human ZNF530 (NP_065931.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348327
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.