Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF534 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541248
Description
Peptide sequence: VFRVEAWRAS GGAEEPTGPN FVGNDSPLPS PLALLPFPSV SASLGDAKRA Sequence homology: Human: 100%.
ZNF534 contains 1 KRAB domain. The exact function of ZNF534 is not known.
Specifications
| ZNF534 | |
| Polyclonal | |
| Unconjugated | |
| ZNF534 | |
| KRAB domain only 3; KRAB domain only protein 3; KRBO3; Zinc finger protein 534; ZNF534 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 147658 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q76KX8 | |
| ZNF534 | |
| Synthetic peptide directed towards the middle region of human ZNF534. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction