Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF536 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520854
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520854 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520854 Supplier Invitrogen™ Supplier No. PA520854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20854 can be used with blocking peptide PEP-0968.

ZNF536 is a recently identified zinc-finger protein that is expressed primarily in the developing nervous system and the cerebral cortex, hippocampus, and hypothalamus. ZNF536 possess ten zinc fingers and interacts with CtBP1, a corepressor for gene transcription. It is most closely related to transcriptional repressor ZNF219. Overexpression of ZNF536 in embryonic stem cells dramatically reduced the mRNA levels of neuronal marker genes such as Pax6, MAP2, and beta-tubulin III following retinoic acid (RA)-induced differentiation, while depletion of ZNF536 via RNAi resulted in elevated mRNA levels of these genes, indicating its role in inhibiting neuronal cell differentiation. Overexpression of RA receptor a rescues the inhibitory role of ZNF536, suggesting that ZNF536 might inhibit RA response element-mediated transcriptional activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF536
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZNF536
Gene Accession No. O15090, Q8K083
Gene Alias 9630010P11Rik; KIAA0390; mKIAA0390; RGD1563363; Zfp536; Zinc finger protein 536; Znf536
Gene Symbols Zfp536, ZNF536
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the amino terminus of human ZNF536.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243937, 292820, 9745
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.