Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF540 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551936 Supplier Invitrogen™ Supplier No. PA551936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSSEKDIHEI SLSKESIIEK SKTLRLKGSI FRNEWQNKSE FEGQQGLKER SISQKKIVSK KMSTDRKRPS FTLNQRIHNS EKSCDSHLVQ HGKIDSDVKH DCKECGSTFN NVYQLTLHQK IHTGEKSCKC EKCGK Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 35%.

ZNF540 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 17 C2H2-type zinc fingers and 1 KRAB domain. ZNF540 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF540
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF540
Gene Accession No. Q8NDQ6
Gene Alias CTD-3064H18.6; Nbla10512; putative protein product of Nbla10512; zinc finger protein 540; ZNF540
Gene Symbols ZNF540
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF540. Recombinant protein control fragment (Product #RP-99756).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.