Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF541 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585001 Supplier Invitrogen™ Supplier No. PA585001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKKVKVDCDS FLCQNPGEPG LQEAQKAGGL PADASPLFRQ LFLKSQEPLV SHEQMQVFQM ITKSQRIFSH AQV Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 75%.

Component of some chromatin remodeling multiprotein complex that plays a role during spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF541
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF541
Gene Accession No. Q9H0D2
Gene Alias Zfp541; Zinc finger protein 541; ZNF541
Gene Symbols ZNF541
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ZNF541 (Product #RP-106867)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.