Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF544 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551737 Supplier Invitrogen™ Supplier No. PA551737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HQRTHTGEKP YECNLCGKSF SQSSKLITHQ RIHTGEKPYQ CIECGKSFRW NSNLVIHQRI HTGEKPYDCT HCGKSFSQSY QLVAHKRTHT GEKPYECNEC GKAFNRSTQL IRHLQIHTGE KPYKCNQCNK A Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF544 is a zinc finger protein belonging to the Kruppel C2H2-type zinc-finger protein family. It localizes to the nucleus and may play a role in transcriptional regulation. ZNF544 is a 715 amino acid long protein that contains 13 C2H2-type zinc fingers and 1 KRAB domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF544
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF544
Gene Accession No. Q6NX49
Gene Alias Zinc finger protein 544; ZNF544
Gene Symbols ZNF544
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF544. Recombinant protein control fragment (Product #RP-102105).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27300
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.