Learn More
Invitrogen™ ZNF550 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589407
Description
Immunogen sequence: TFTREEWRQL DLAQRTLYRE VMLETCGLLV SLGHRVPKPE LVHLLEHGQE LWIVKRGLSH ATCAGDRAQV HTREPTTYPP VLSERAFLRG SLTLESSTSS DSRLGRARDE EGLLEMQKGK VTPETDLHKE THLGKVSLEG EGLGTDDGLH SRALQEWLSA DVLHECDSQQ PGKDALIHAG T; Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus.
May be involved in transcriptional regulation.
Specifications
ZNF550 | |
Polyclonal | |
Unconjugated | |
Zinc finger protein 550; ZNF550 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human ZNF550 (NP_0012640191). | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.92 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.