Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF551 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565208 Supplier Invitrogen™ Supplier No. PA565208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFAHVTSLGYC HGMENEAIAS EQSVSIQVRT SKGNTPTQKT HLSEIKMCVP VL Highest antigen sequence identity to the following orthologs - mouse 42%, rat 40%.

Located on chromosome 19, ZNF551 encodes a protein that is part of the zinc finger protein family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF551
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF551
Gene Accession No. Q7Z340
Gene Alias KOX 23 protein (56 AA); KOX23; Zinc finger protein 551; zinc finger protein KOX23; ZNF551
Gene Symbols ZNF551
Host Species Rabbit
Immunogen Recombinant Human ZNF551. Recombinant protein control fragment (Product #RP-105946).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.