Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF554 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530907 Supplier Invitrogen™ Supplier No. PA530907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZNF554, or Zinc Finger Protein 554, is a member of the Kruppel-associated box domain-zinc finger proteins (KRAB-ZNF) subfamily, residing on chromosome 19p13. This region is characterized by a concentration of heterochromatin proteins, which indicates tightly regulated gene expression. ZNF554 is implicated in various cellular processes including transcription regulation and apoptosis. Dysregulation of ZNF554 has been linked to several diseases; notably, its decreased expression is associated with poor prognosis in gliomas, specifically grades III and IV astrocytomas, where it affects the PI3K-Akt signaling pathway. It also plays a role in placental development, with dysfunction contributing to conditions such as fetal growth restriction (FGR). Additionally, ZNF554 transcription is influenced by factors like ZEB1, ZEB2, and hypoxic conditions, with neighboring hypoxia-response elements showing hypermethylation in preeclampsia. This gene's expression can significantly alter the transcriptional landscape, with overexpression leading to the differential regulation of hundreds of genes, underscoring its importance in cellular regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF554
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNF554
Gene Accession No. Q86TJ5
Gene Alias FLJ34817; Zinc finger protein 554; ZNF554
Gene Symbols ZNF554
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF554. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115196
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.