Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF555 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562883 Supplier Invitrogen™ Supplier No. PA562883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEALSQIPHL NLYKKIPPGV KQYEYNTYGK VFMHRRTSLK SPITVHTGHK PY Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 46%.

ZNF555 contains 15 C2H2-type zinc fingers and 1 KRAB domain. It belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF555
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF555
Gene Accession No. Q8NEP9
Gene Alias MGC26707; Zinc finger protein 555; ZNF555
Gene Symbols ZNF555
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF555. Recombinant protein control fragment (Product #RP-101563).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148254
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.