Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF556 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536014
Description
Recommended positive controls: RD. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in transcriptional regulation.
Specifications
| ZNF556 | |
| Polyclonal | |
| Unconjugated | |
| ZNF556 | |
| Zinc finger protein 556; ZNF556 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80032 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9HAH1 | |
| ZNF556 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF556. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction