Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF557 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541175
Description
Peptide sequence: GKAFGTRSSL SSHYSIHTGE YPYECHDCGR TFRRRSNLTQ HIRTHTGEKP Sequence homology: Cow: 77%; Dog: 77%; Guinea Pig: 77%; Horse: 77%; Human: 100%; Mouse: 77%; Rat: 77%.
ZNF557 is a new candidate transcription factor.
Specifications
| ZNF557 | |
| Polyclonal | |
| Unconjugated | |
| ZNF557 | |
| CTB-25J19.9; zinc finger protein 557; ZNF557 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79230 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N988 | |
| ZNF557 | |
| Synthetic peptide directed towards the middle region of human ZNF557. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction