Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF557 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120646 Supplier Invitrogen™ Supplier No. PA5120646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, A-549, 293T, Mouse lung, Mouse brain, Rat lung. Immunogen sequence: MAAVVLPPTA ALSSLFPASQ REGHTEGGEL VNELLKSWLK GLVTFEDVAV EFTQEEWALL DPAQRTLYRD VMLENCRNLA SLGNQVDKPR LISQLEQEDK VMTEERGILS GTCPDVENPF KAKGLTPKLH VFRKEQSRNM KMERNHLGAT.

ZNF557 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF557
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF557
Gene Accession No. Q8N988
Gene Alias CTB-25J19.9; zinc finger protein 557; ZNF557
Gene Symbols ZNF557
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human ZNF557 (NP_077317.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79230
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.