Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF558 Monoclonal Antibody (OTI2C4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527410

Catalog No. PIMA527410


Only null left
Add to Cart

Description

Description

ZNF558 Monoclonal Antibody for Western Blot, IHC (P)

ZNF558 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF558
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96NG5
ZNF558
Full length human recombinant protein of ZNF558 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI2C4
Unconjugated
ZNF558
Zinc finger protein 558; ZNF558
Mouse
Affinity Chromatography
RUO
148156
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.