Learn More
ZNF558 Mouse anti-Human, Clone: OTI2C4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811169

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF558 may be involved in transcriptional regulation.Specifications
ZNF558 | |
Monoclonal | |
Unconjugated | |
ZNF558 | |
zinc finger protein 558 | |
Mouse | |
Affinity Chromatography | |
RUO | |
148156 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96NG5 | |
ZNF558 | |
Full length human recombit protein of human ZNF558 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.