Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF559 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541184
Description
Peptide sequence: LPSSSHLREC VRIYGGERPY THKEYVETFS HSTALFVHMQ TQDGEKFYEC Sequence homology: Human: 100%.
ZNF559 may be involved in transcriptional regulation.
Specifications
| ZNF559 | |
| Polyclonal | |
| Unconjugated | |
| ZNF559 | |
| NBLA00121; putative protein product of Nbla00121; zinc finger protein 559; ZNF559 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 84527 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BR84 | |
| ZNF559 | |
| Synthetic peptide directed towards the N-terminal of human ZNF559 (aa 194-243). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction