Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF561 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568718
Description
This target displays homology in the following species: Cow: 77%; Dog: 77%; Guinea Pig: 77%; Horse: 77%; Human: 100%; Mouse: 85%; Pig: 85%; Rabbit: 85%; Rat: 77%.
May be involved in transcriptional regulation.
Specifications
ZNF561 | |
Polyclonal | |
Unconjugated | |
ZNF561 | |
Zinc finger protein 561; ZNF561 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
93134 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N587 | |
ZNF561 | |
synthetic peptide directed towards the C terminal region of human ZNF561. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction