Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF562 Antibody (PCRP-ZNF562-1A1), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314150AF647
This item is not returnable.
View return policy
Description
ZNF562 Monoclonal antibody specifically detects ZNF562 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ZNF562 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
54811 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ZNF562-1A1 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
FLJ20079, MGC126160, zinc finger protein 562 | |
Recombinant full-length human ZNF562 protein (Uniprot: Q6V9R5) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction