Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF562 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | ZNF562 |
---|---|
Applications | Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
ZNF562 Polyclonal specifically detects ZNF562 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZNF562 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
54811 | |
IgG | |
Affinity purified |
Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
FLJ20079, MGC126160, zinc finger protein 562 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human ZNF562 (NP_001123503.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title