Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF563 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543634
Description
Peptide sequence: ETIRNLDCIR MIWEEQNTED QYKNPRRNLR CHMVERFSES KDSSQCGETF Sequence homology: Human: 100%; Mouse: 82%; Pig: 80%.
ZNF563 may be involved in transcriptional regulation.
Specifications
ZNF563 | |
Polyclonal | |
Unconjugated | |
ZNF563 | |
Zinc finger protein 563; ZNF563 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
147837 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TA94 | |
ZNF563 | |
Synthetic peptide directed towards the N-terminal of human ZNF563 (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction