Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF568 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569061
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 100%; Sheep: 77%.
ZNF568 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
ZNF568 | |
Polyclonal | |
Unconjugated | |
ZNF568 | |
LOC100061744; LOW QUALITY PROTEIN: zinc finger protein 568; zinc finger protein 568; ZNF568 | |
Rabbit | |
Affinity chromatography | |
RUO | |
374900 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3ZCX4 | |
ZNF568 | |
synthetic peptide directed towards the N terminal region of human ZNF568. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction