Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF568 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982055
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2055 Supplier Bioss Supplier No. BS12228R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF568 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.

Specifications

Antigen ZNF568
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF568
Gene Accession No. Q3ZCX4
Gene Alias LOW QUALITY PROTEIN: zinc finger protein 568; zinc finger protein 382; Zinc finger protein 568; ZNF382; ZNF568
Gene Symbols ZNF568
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF568.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374900
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.