Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF569 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562713 Supplier Invitrogen™ Supplier No. PA562713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIWGVDEHQK NQDRLLRQVE VKFQKTLTEE KGNECQKKFA NVFPLNSDFF PSRHNLYEYD LFGKCLEHNF DCHNNVKCLM RKEHCEYNEP VKSYGNSSSH FVIT Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 44%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF569
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF569
Gene Accession No. Q5MCW4
Gene Alias 2810054M15Rik; DNA-binding protein; KRAB8; LOW QUALITY PROTEIN: zinc finger protein 569; Mszf21; mszf77; Unknown (protein for IMAGE:7986680); ZAP1; Zfp66; Zfp74; Zfp-74; zinc finger protein 568; zinc finger protein 569; Zinc finger protein 74; ZNF; Znf569
Gene Symbols ZNF569
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF569. Recombinant protein control fragment (Product #RP-92441).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148266
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.