Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF57 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25671025UL
Description
ZNF57 Polyclonal specifically detects ZNF57 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ZNF57 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
Zinc finger protein 424ZNF424, zinc finger protein 57 | |
Rabbit | |
Affinity Purified | |
RUO | |
126295 | |
Human | |
Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
ZNF57 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TGQKSHECQSYSKAFSCQVILSKTSESTH | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction