Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541684 Supplier Invitrogen™ Supplier No. PA541684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MDSVVFEDVA VDFTLEEWAL LDSAQRDLYR DVMLETFRNL ASVDDGTQFK Sequence homology: Cow: 100%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 100%.

The function of ZNF57 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF57
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF57
Gene Accession No. Q68EA5
Gene Alias Zinc finger protein 424; zinc finger protein 57; ZNF424; ZNF57
Gene Symbols ZNF57
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF57 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126295
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.