Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF574 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10097825AF405
Description
ZNF574 Polyclonal antibody specifically detects ZNF574 in Human samples. It is validated for Western BlotSpecifications
ZNF574 | |
Polyclonal | |
Western Blot | |
FLJ22059, FP972, zinc finger protein 574 | |
The immunogen recognized by this antibody maps to a region between residue 846 and 896 of human zinc finger protein 574 using the numbering given in entry NP_073589.4 (GeneID 64763). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
64763 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction