Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF577 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561310 Supplier Invitrogen™ Supplier No. PA561310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGIAEGAAHS QICPGFVIQS RRYAGKDSDA FGGYGRSCLH IKRDKTLTGV KYHRCVKPSS PKSQLNDLQK ICAGGKPH Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 29%.

ZNF577 is a candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF577
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF577
Gene Accession No. Q9BSK1
Gene Alias Zinc finger protein 577; ZNF577
Gene Symbols ZNF577
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF577. Recombinant protein control fragment (Product #RP-98518).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.