Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF579 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568719
Description
This target displays homology in the following species: Human: 100%; Mouse: 86%; Rabbit: 79%; Rat: 86%.
May be involved in transcriptional regulation.
Specifications
ZNF579 | |
Polyclonal | |
Unconjugated | |
ZNF579 | |
1110003A17Rik; FLJ35453; Zfp579; Zinc finger protein 579; ZNF579 | |
Rabbit | |
Affinity chromatography | |
RUO | |
163033 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NAF0 | |
ZNF579 | |
synthetic peptide directed towards the C terminal region of human ZNF579. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction