Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF580 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541574
Description
Peptide sequence: KAEGPSSTPS SAAGPRPPRL GRHLLIDANG VPYTYTVQLE EEPRGPPQRE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rat: 93%.
ZNF580 may be involved in transcriptional regulation.
Specifications
ZNF580 | |
Polyclonal | |
Unconjugated | |
ZNF580 | |
1500015O20Rik; AI838225; LDL induced EC protein; LDL-induced EC protein; Zfp580; Zinc finger protein 580; zinc finger protein 865; zinc finger protein 580; ZNF580; ZNF865 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51157 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UK33 | |
ZNF580 | |
Synthetic peptide directed towards the N-terminal of human ZNF580 (aa 31-80). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction