Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF582 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541208 Supplier Invitrogen™ Supplier No. PA541208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SGGLCPVLES RYDTKELFPK QHVYEVESPQ WEIMESLTSY GLECSSFQDD Sequence homology: Horse: 85%; Human: 100%; Pig: 79%.

ZNF582 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 10 C2H2-type zinc fingers and 1 KRAB domain. ZNF582 ay be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF582
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF582
Gene Accession No. Q96NG8
Gene Alias Zinc finger protein 582; ZNF582
Gene Symbols ZNF582
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF582 (aa 72-121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147948
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.