Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF583 Monoclonal Antibody (OTI4D2)
GREENER_CHOICE

Catalog No. PIMA527319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527319 Supplier Thermo Scientific Supplier No. MA527319
Add to Cart
Add to Cart

ZNF583 Monoclonal Antibody (OTI4D2)

ZNF583 Monoclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF583
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF583
Gene Accession No. Q96ND8
Gene Alias BC056221; LOC101788406; LOW QUALITY PROTEIN: zinc finger protein 583; RGD1562044; Zfp583; Zinc finger protein 583; zinc finger protein L3-5; Znf583
Gene Symbols ZNF583
Host Species Mouse
Immunogen Full length human recombinant protein of ZNF583 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147949
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.