Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF583 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554890
Description
Immunogen sequence: EPWMVKKEGT RGPCPDWEYV FKNSEFSSKQ ETYEESSKVV TVGARHLSYS LDYPSLREDC QSEDWYKNQL GSQEVH Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 55%.
ZNF583 may be involved in transcriptional regulation.
Specifications
| ZNF583 | |
| Polyclonal | |
| Unconjugated | |
| ZNF583 | |
| BC056221; LOC101788406; LOW QUALITY PROTEIN: zinc finger protein 583; RGD1562044; Zfp583; Zinc finger protein 583; zinc finger protein L3-5; Znf583 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 147949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Lot-Specific | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96ND8 | |
| ZNF583 | |
| Recombinant protein corresponding to Human ZNF583. Recombinant protein control fragment (Product #RP-93651). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction