Learn More
Invitrogen™ ZNF583 Polyclonal Antibody

Description
Immunogen sequence: LSQLIITHKE ILPEVQNKEY NKSWQTFHQD TIFDIQQSFP TKEKAHKHEP QKKSYRKKSV EMKHRKVYV Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 59%.
Specifications
Specifications
| Antigen | ZNF583 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF583 |
| Gene Accession No. | Q96ND8 |
| Gene Alias | BC056221; LOC101788406; LOW QUALITY PROTEIN: zinc finger protein 583; RGD1562044; Zfp583; Zinc finger protein 583; zinc finger protein L3-5; Znf583 |
| Gene Symbols | ZNF583 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.