Learn More
Invitrogen™ ZNF587 Monoclonal Antibody (3A3)

Description
Peptide Sequence: MAVSSQQGEI MESRIFFQGS HAHFPTCMNV DTAATVLAVN VNLASNHCSQ GNVPIRRRLS GTLILTGRWD ILRDPEAGCH LLNFPEGCLE SVSSHSELFF LLWLTKNMEP HKVHCNSFIF VK.
Specifications
Specifications
| Antigen | ZNF587 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3A3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF587 |
| Gene Accession No. | Q96SQ5 |
| Gene Alias | ZF6; Zinc finger protein 587; zinc finger protein zfp6; ZNF587 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.