Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF592 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074658AF405
Description
ZNF592 Polyclonal antibody specifically detects ZNF592 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ZNF592 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CAMOS, KIAA0211MGC138437, MGC138439, SCAR5, spinocerebellar ataxia, autosomal recessive 5, zinc finger protein 592 | |
The immunogen recognized by this antibody maps to a region between residue 950 and 1000 of human zinc finger protein 592 using the numbering given in entry NP_055445.2 (GeneID 9640). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9640 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction