Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF592 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554467 Supplier Invitrogen™ Supplier No. PA554467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LARELGTCSS VPPRQRLKPA HSKLSSCVAA LVALQAKRVA SVTKEDQPGH TKDLSGPTKE SSKGSPKMPK SPKSPRSPLE ATRKSIKPSD SPRSICSDSS SKGSPSVAAS S Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.

This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia. [provided by RefSeq, Jan 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF592
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF592
Gene Accession No. Q92610
Gene Alias 8430405N24; A730014M16Rik; CAMOS; KIAA0211; mKIAA0211; SCAR5; Zfp592; Zfp-592; Zinc finger protein 592; ZNF592
Gene Symbols ZNF592
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF592. Recombinant protein control fragment (Product #RP-91690).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9640
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.