Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF593 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565865 Supplier Invitrogen™ Supplier No. PA565865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSTNLKTHFRS KDHKKRLKQL SVEPYSQEEA ERAAGMGSYV PPRRLAVPTE VSTEVPEMDT ST Highest antigen sequence identity to the following orthologs - mouse 90%, rat 92%.

Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF593
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF593
Gene Accession No. O00488
Gene Alias 3110024A21Rik; AV062409; E130106C14Rik; Zfp593; Zinc Finger 593; Zinc finger protein 593; zinc finger protein T86; ZNF593; ZT86
Gene Symbols ZNF593
Host Species Rabbit
Immunogen Recombinant Human ZNF593. Recombinant protein control fragment (Product #RP-106536).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51042
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.