Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF593 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571626
Description
ZNF593 Polyclonal Antibody for Western Blot
Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.
Specifications
ZNF593 | |
Polyclonal | |
Unconjugated | |
ZNF593 | |
3110024A21Rik; AV062409; E130106C14Rik; Zfp593; Zinc Finger 593; Zinc finger protein 593; zinc finger protein T86; ZNF593; ZT86 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51042, 68040 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O00488, Q9DB42 | |
Zfp593, ZNF593 | |
KLH conjugated synthetic peptide between 62-88 amino acids from the Central region of human ZNF593. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction