Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF594 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321372100UL
Description
ZNF594 Polyclonal antibody specifically detects ZNF594 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ZNF594 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
KIAA1871, Zinc Finger Protein 594, Zinc Finger Protein HZF18 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: IREMHIIPQKAIVGEIGHGCNEGEKILSAGESSHRYEVSGQNFKQKSGLTEHQK | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
84622 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction