Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF594 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584448 Supplier Invitrogen™ Supplier No. PA584448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IREMHIIPQK AIVGEIGHGC NEGEKILSAG ESSHRYEVSG QNFKQKSGLT EHQK Highest antigen sequence indentity to the following orthologs: Mouse - 33%, Rat - 31%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF594
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF594
Gene Accession No. Q96JF6
Gene Alias KIAA1871; Zinc finger protein 594; zinc finger protein HZF18; ZNF594
Gene Symbols ZNF594
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF594. Recombinant protein control fragment (Product #RP-108443).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.